Order Entry
France
ContactUsLinkComponent
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Certificates
undefined
Fehling's reagent I (copper(II) sulphate solution) for qualitative determination of reducing sugars
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Clear blue Solution
  • Solution contains:
    69.2 g CuSO4.5H2O/l H2O