Order Entry
France
ContactUsLinkComponent
Sodium hypochlorite (0.5% Cl₂) in aqueous solution
Sodium hypochlorite (0.5% Cl₂) in aqueous solution
Catalog # PRLS85471.360
Supplier:  PROLABO CMP
CAS Number:  
Sodium hypochlorite (0.5% Cl₂) in aqueous solution
Catalog # PRLS85471.360
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :74.44 g/mol
Storage Temperature:Refrigerator
EINECS:231-668-3
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120