Lanolin, anhydrous USP
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8006-54-0
:
MDL Number:MFCD00081740
Merck Index:12,05371
Melting Pt:38…40 °C
Density:0,932 to 0.945 g/cm³ (15 °C)
Flash Pt:209 °C (closed cup)
Storage Temperature:Ambient
EINECS:232-348-6
Specification Test Results
- Appearance:Yellow Wax
- Infrared spectrum:Conforms
- Melting point:38 to 44 °C
- Water:≤0.25 %
- Additional info:Water-soluble acids and alkalies: passes test
- Antioxidant:≤0.02 % (BHT)
- Residue after ignition:≤0.1 %
- Acidity:≤2.0 ml (ml 0.1 N NaOH)
- Alkalinity:Passes test
- Chloride (Cl):≤0.035 %
- Iodine value:18 to 36
- Ammonia (NH3):Passes test
Specification Test Results
Related Information
Frequently Bought Together