Order Entry
France
ContactUsLinkComponent
Solvent ASTM D4739, Toluene : Propan-2-ol : Chloroform : water (10:10:10:0.3) (v/v/v/v) , Toluene/ Propan-2-ol/ Chloroform/ water, (10/10/10/0,3 v/v), VOLUSOL®
Solvent ASTM D4739, Toluene : Propan-2-ol : Chloroform : water (10:10:10:0.3) (v/v/v/v) , Toluene/ Propan-2-ol/ Chloroform/ water, (10/10/10/0,3 v/v), VOLUSOL®
Catalog # 5740.2500
Supplier:  VWR Chemicals
CAS Number:  
undefined
Solvent ASTM D4739, Toluene : Propan-2-ol : Chloroform : water (10:10:10:0.3) (v/v/v/v) , Toluene/ Propan-2-ol/ Chloroform/ water, (10/10/10/0,3 v/v), VOLUSOL®
Catalog # 5740.2500
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identification:
    Passes test
  • Density (20/4):
    1.03 - 1.046

Specification Test Results

About this item

 :
UN:1993
ADR:3,II
 :GHS02
 :GHS08
 :GHS07