Benzethonium chloride 0.004 mol/l, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride
CAS Number:121-54-0
:
Formula:C₂₇H₄₂ClNO₂
MDL Number:MFCD00011742
:448.09 g/mol
EINECS:204-479-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.00399 - 0.00401 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500)
Packaging Information
spec_VWRC30497.293 spec_VWRC30497.293 spec_VWRC30497.327 spec_VWRC30497.327
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together