Order Entry
France
ContactUsLinkComponent
Patton-Reeders reagent (Calconcarboxylic acid) analytical reagent
Irritant
Patton-Reeders reagent (Calconcarboxylic acid) analytical reagent
Catalog # 20326.136
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Patton-Reeders reagent (Calconcarboxylic acid) analytical reagent
Catalog # 20326.136
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Suited for calcium reagent:
    Passes test

Specification Test Results

About this item

 :
 :438.42 g/mol
Melting Pt:300 °C
EINECS:223-117-0
 :GHS07
Storage Temperature:Ambient
CAS Number:3737-95-9
Formula:C₂₁H₁₄N₂O₇S
MDL Number:MFCD00004078