Order Entry
France
ContactUsLinkComponent
PAH Mix 1, EPA 610
PAH Mix 1, EPA 610
Catalog # 124812X
Supplier:  VWR Chemicals
CAS Number:  
undefined
PAH Mix 1, EPA 610
Catalog # 124812X
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Vial
  • Pk:
    1 mL
  • Description:
    PAH Mix 1, EPA 610
Specifications
About this item
Multi-component standard with 0,1 mg/ml of each component in methanol.
16 components: Acenaphthene Chrysene; Acenaphthylene Dibenz[a,h]anthracene; Anthracene Fluoranthene; Benzo[a]anthracene Fluorene; Benzo[a]pyrene Indeno[1,2,3-cd]pyrene; Benzo[b]fluoranthene Naphthalene; Benzo[g,h,i]perylene Phenanthrene; Benzo[k]fluoranthene Pyrene.