Order Entry
France
ContactUsLinkComponent
Smart probes, VAC kit
Smart probes, VAC kit
Catalog # TEST0563.0003.10
Supplier:  TESTO
undefined
Smart probes, VAC kit
Catalog # TEST0563.0003.10
Supplier:  TESTO
Specifications
  • Range:
    0 - 30 m/s [testo 405i], 0,4 - 30 m/s [testo 410i], 0 - 100 %RH [testo 605i], -30...250 °C [testo 805i]
  • Description:
    testo Smart Probes VAC kit for servicing on ventilation and air conditioning systems
  • Seq:
    03
  • Model:
    VAC kit (testo 405i, testo 410i, testo 605i, testo 805i)
  • Batteries:
    3 micro batteries AAA
  • Sensor:
    Hot wire [testo 405i], Vane [testo 410i], Humidity – capacitive [testo 605i], Infrared [testo 805i]
  • Pk:
    1 KIT
Specifications
About this item
Testo 405i thermal anemometer, testo 410i vane anemometer, testo 605i thermohygrometer, and testo 805i infrared thermometer in the testo Smart Case.
  • Compact professional measuring instruments from the Testo Smart Probes series for use with smartphones/tablets
  • For the measurement of air and surface temperature, humidity, air velocity and volume flow
  • Measurement data displayed as a table or graphic
  • Measurement data analyzed and sent via testo Smart Probes App
  • Handy testo Smart Case transport bag
Application-specific menus: volume flow measurement in ducts and at outlets, identification of mould risk, easy image documentation including IR temperature measuring value and measurement spot marking.
The compact VAC kit includes four App-controlled measuring instruments for the most important measuring tasks performed by VAC engineers: the testo 405i thermal anemometer, the testo 410i vane anemometer, the testo 605i thermohygrometer and the testo 805i infrared thermometer. In conjunction with a smartphone or tablet, it can be used to measure air and surface temperatures, air humidity, air velocities and volume flows in, on and around air conditioning plants and systems. Users can read off their measuring values conveniently via the testo Smart Probes App installed on the smartphone/tablet.
In the App, measurement parameters (such as humidity or temperature) can be deleted, added or their order altered with just one click. It is also possible to change the displayed measurement parameters just as quickly. In addition to this, the App offers application-specific menus, including for the identification of mould risks; automatic calculation of volume flow at outlets/in ducts by easy parameterization of the outlet/duct cross-section (dimensions and geometry). Finally, the measurement data report can be e-mailed directly as a pdf or Excel file. In the handy testo Smart Case, the measuring instruments can be conveniently transported and are always to hand when they are needed.
testo Smart Probes App turns your smartphone/tablet into the display for up to 6 Testo Smart Probes at the same time. Both the operation of the measuring instruments and the display of the readings are achieved by Bluetooth® via the testo Smart Probes App on your smartphone or tablet – irrespective of the measuring location. In addition, you can use the App to create measurement reports, add photos and comments to these and send them by e-mail. For iOS and Android.
Ordering information: Requires iOS 8.3 or newer/Android 4.3 or newer, and mobile terminal device with Bluetooth® 4.0.
Delivery: Kit consists of testo 405i, testo 410i, testo 605i, testo 805i, testo Smart Case (VAC), batteries, and calibration protocol.