Order Entry
France
ContactUsLinkComponent
Rat Recombinant IL-3 B (from E. coli)
Rat Recombinant IL-3 B (from E. coli)
Certificates
undefined
Rat Recombinant IL-3 B (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1,0×10⁴ units/mg
  • Gene ID:
    P04823
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    MCGF|P-cell stimulating factor|HCGF|Multi-CSF
  • Protein/peptide name:
    IL-3 B (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    16,3 kDa
  • Sequence:
    MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together