Order Entry
France
ContactUsLinkComponent
Oleic acid, Extra pure meets analytical specification of Ph.Eur., Fluka™
Irritant
Oleic acid, Extra pure meets analytical specification of Ph.Eur., Fluka™
Catalog # HONC27728H-25KG
CAS Number:  
undefined
Irritant
Oleic acid, Extra pure meets analytical specification of Ph.Eur., Fluka™
Catalog # HONC27728H-25KG
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay (GC):
    65.0 - 88.0 %
  • Density (D 20/20):
    0.889 - 0.895
  • Ash:
    Max. 0.1 %
  • Iodine value:
    89 - 105
  • Peroxide value:
    Max. 10.0
  • Acid value:
    195 - 204
  • Appearance of the substance:
    complying
  • Identity:
    complying
  • Fatty acid composition:
    complying
  • Margaric acid (GC):
    Max. 0.2 %
  • Assay of myristic acid (GC):
    Max. 5.0 %
  • Assay of palmitic acid (GC):
    Max. 16.0 %
  • Assay of palmitoleic acid (GC):
    Max. 8.0 %
  • Assay of stearic acid (GC):
    Max. 6.0 %
  • Assay of oleic acid (GC):
    65.0 - 88.0 %
  • Assay of linoleic acid (GC):
    Max. 18.0 %
  • Assay of linolenic acid (GC):
    Max. 4.0 %
  • Fatty acids w. 1 chain >C18:
    Max. 4.0 %
  • Residual solvents:
    complying

Specification Test Results

About this item

 :
Boiling Pt:360 °C (1.013 hPa)
Melting Pt:13-14 °C
Melting Pt:13…14 °C
Density:0.9 g/cm3 (20 °C)
Flash Pt:113 °C
 :282.47 g/mol
Storage Temperature:Ambient
EINECS:204-007-1
 :GHS07
CAS Number:112-80-1
Formula:C₁₈H₃₄O₂
MDL Number:MFCD00064242