Order Entry
France
ContactUsLinkComponent
COVID-19 IgG/IgM rapid test cassette
COVID-19 IgG/IgM rapid test cassette
Catalog # 720-2967
Supplier:  SURESCREEN
CAS Number:  
undefined
COVID-19 IgG/IgM rapid test cassette
Catalog # 720-2967
Supplier:  SURESCREEN
CAS Number:  
Specifications
  • Pk:
    25
  • Description:
    Rapid test cassette for COVID-19 coronavirus
Specifications
About this item
Rapid test for the qualitative detection of antibodies (IgG and IgM) against SARS-CoV-2 in whole blood, serum or plasma. For professional in vitro diagnostic use only.
The COVID-19 IgG/IgM Rapid Test Cassette (Whole Blood/Serum/Plasma) is a rapid chromatographic immunoassay for the qualitative detection of IgG and IgM antibodies against SARS-CoV-2 in whole blood, serum or plasma. human plasma, intended to aid in the diagnosis of primary and secondary infections with SARS-CoV-2.
Caution: Not for self-testing. The test is intended for professional in vitro diagnostic use by health care professionals only.