Order Entry
France
ContactUsLinkComponent
Piperidine ≥99.0% (by GC), GPR RECTAPUR®
Flammable
Toxic
Corrosive
Piperidine ≥99.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Toxic
Corrosive
Piperidine ≥99.0% (by GC), GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance :
    Clear liquid
  • Assay:
    Min. 99.0 %
  • Density (20/4):
    0.861 - 0.863
  • Water :
    Max. 0.5 %
  • Identity (IR):
    Passes test

Specification Test Results

Frequently Bought Together