Order Entry
France
ContactUsLinkComponent
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Certificates
undefined
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Certificates

Specification Test Results

  • Identity (GC):
    complying
  • Assay (GC):
    99.0 - 99.4 %
  • Boiling range:
    60 - 62 °C
  • Density (D 20/20):
    1.476 - 1.483
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as HCl):
    Max. 0.00005 %
  • Acid and Phosgene:
    complying
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.000005 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • Chloride (Cl):
    Max. 0.0001 %
  • Free chlorine (Cl):
    Max. 0.00001 %
  • Aldehydes, Ketones (as CH₃CocH₃):
    Max. 0.005 %
  • Carbonyl compounds (as CO):
    Max. 0.005 %
  • Dichloromethane (GC):
    Max. 0.01 %
  • Ethanol (GC):
    0.6 - 1.0 Gew.%
  • Tetrachloroethene (GC):
    Max. 0.01 %
  • Tetrachloromethane (GC):
    Max. 0.01 %
  • Trichloroethene (GC):
    Max. 0.01 %
  • Other Impurities (GC):
    Max. 0.1 %
  • Reaction against H₂SO₄:
    complying
  • Appearance of the substance:
    complying
  • Suitability for determinations with dithizone (ISO):
    complying

Specification Test Results

Related Information

Frequently Bought Together