3-Ferrocenylpropionic anhydride ≥98.0% (C basis), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 76737-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:498.17 g/mol
:GHS05
CAS Number:132098-76-1
Formula:C₂₆H₂₆Fe₂O₃
MDL Number:MFCD18071429