Order Entry
France
ContactUsLinkComponent
N-Succinimidyl 4-(dimethylamino)benzoate ≥98.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
N-Succinimidyl 4-(dimethylamino)benzoate ≥98.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 61224-500MG
Supplier:  Merck
CAS Number:  
undefined
N-Succinimidyl 4-(dimethylamino)benzoate ≥98.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 61224-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :262.26 g/mol
CAS Number:58068-85-2
Formula:C₁₃H₁₄N₂O₄
MDL Number:MFCD16875522