Calcimycin (A23187), Selectophore™, function tested ionophore, Supelco®
Catalog # 21186-25MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:523.63 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:52665-69-7
Formula:C₂₉H₃₇N₃O₆
MDL Number:MFCD00151202