Order Entry
France
ContactUsLinkComponent
Reserpine 5 mg/l in 2-propanol 50% (v/v) analytical standard, for LC-MS, Supelco®
Flammable
Irritant
Reserpine 5 mg/l in 2-propanol 50% (v/v) analytical standard, for LC-MS, Supelco®
Catalog # 43530-4.5ML-F
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Reserpine 5 mg/l in 2-propanol 50% (v/v) analytical standard, for LC-MS, Supelco®
Catalog # 43530-4.5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    4,5 mL
  • Seq:
    2205

Specifications

About this item

 :
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :608.69 g/mol
UN:1219
 :GHS02
 :GHS07
CAS Number:50-55-5
Formula:C₃₃H₄₀N₂O₉
MDL Number:MFCD00005091