


n-Octane ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Catalog # 74821-500ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Identity (NMR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.397 - 1.399
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.02 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Explosion Limit:6.5 %
Specification Test Results
About this item
:
:114.23 g/mol
Merck Index:13,06782
Boiling Pt:6.5 %(V)
Melting Pt:-57 °C
Melting Pt:–57 °C
Density:0.703 g/cm³ (25 °C)
Flash Pt:13 °C
EINECS:203-892-1
UN:1262
ADR:3,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:111-65-9
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556