0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Standards
Organic Standards
Gamithromycin, analytical standard, VETRANAL™, Supelco®
Gamithromycin, analytical standard, VETRANAL™, Supelco®
32161-25MG
:
Merck
:
Gamithromycin, analytical standard, VETRANAL™, Supelco®
32161-25MG
:
Merck
:
Pk:
25 mg
Standards:
analytical standard
CAS No.:
145435-72-9
Description:
Gamithromycin
Description:
Gamithromycin