Order Entry
France
ContactUsLinkComponent
PAS (Periodic Acid Schiff) staining kit, Sigma-Aldrich®
PAS (Periodic Acid Schiff) staining kit, Sigma-Aldrich®
  1.01646.0001
 :  Merck
 :  
undefined
PAS (Periodic Acid Schiff) staining kit, Sigma-Aldrich®
  1.01646.0001
 :  Merck
 :  
  • Pk:
    400 Tests
  • Seq:
    0810
  • Pk Info:
    2 x 500 ml (Preparation Capacity 2500 - 3000 slides)
  • Suitability for microscopy:
    passes test
  • nuclei:
    Blue
  • acid mucosubstances:
    light blue
  • polysaccharides:
    purple
  • neutral mucopolysaccharides:
    purple
 :
 :Ambient