Order Entry
France
ContactUsLinkComponent
Phytochemical standard, Tanshinone I, analytical standard, Supelco®
Phytochemical standard, Tanshinone I, analytical standard, Supelco®
Catalog # 80714-10MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, Tanshinone I, analytical standard, Supelco®
Catalog # 80714-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    568-73-0
  • Description:
    Tanshinone I, ≥98,0% (HPLC)
  • Formula:
    C₁₈H₁₂O₃
  • Storage temperature:
    2...8°C
  • Description:
    Tanshinone I
  • Form:
    Neat

Specifications

About this item

Phenanthrenequinone constituent of Chinese medicinal herb Danshen (Salvia miltiorrhiza). Modulates or prevents breast cancer metastasis by regulating adhesion molecules. Anti-inflammatory.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.