Order Entry
France
ContactUsLinkComponent
 
Organic reference standard, Sudan IV-d6, analytical standard, Supelco®
Catalog # 34161-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sudan IV-d6, analytical standard, Supelco®
Catalog # 34161-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1014689-18-9
  • Description:
    Sudan IV-d6
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₆H₁₂N₂O
  • Description:
    Sudan IV-d6
  • Form:
    Neat

Specifications

About this item