Phytochemical standard, Sinensetin, analytical standard, Supelco®
Catalog # 89392-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:2306-27-6
- Description:Sinensetin
- Formula:C₂₀H₂₀O₇
- Storage temperature:2...8 °C
- Description:Sinensetin
- Form:Neat