Order Entry
France
ContactUsLinkComponent
Organic reference standard, Paraben target analyte mix solution, analytical standard, Supelco®
Organic reference standard, Paraben target analyte mix solution, analytical standard, Supelco®
Catalog # 32127-1ML
Supplier:  Merck
CAS Number:  
Organic reference standard, Paraben target analyte mix solution, analytical standard, Supelco®
Catalog # 32127-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in acetone
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • Description:
    Paraben target analyte mix solution
  • Storage temperature:
    –20 °C
  • Description:
    Paraben target analyte mix solution

Specifications

About this item