Order Entry
France
ContactUsLinkComponent
Phytochemical standard, Neohesperidin, analytical standard, Supelco®
Phytochemical standard, Neohesperidin, analytical standard, Supelco®
Catalog # 40927-10MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, Neohesperidin, analytical standard, Supelco®
Catalog # 40927-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    13241-33-3
  • Description:
    Neohesperidin, ≥95,0% (HPLC)
  • Formula:
    C₂₈H₃₄O₁₅
  • Storage temperature:
    2…8 °C
  • Description:
    Neohesperidin
  • Form:
    Neat

Specifications

About this item

Flavanone glycoside with neuroprotective and antioxidant properties. Does not inhibit oral carcinogenesis in a rat model unlike other citrus flavanones, but does exert a protective effect against gastritis and gastric lesions.