Order Entry
France
ContactUsLinkComponent
 
Phytochemical standard, Glabridin, analytical standard, Supelco®
  53633-10MG
 :  Merck
 :  
undefined
Phytochemical standard, Glabridin, analytical standard, Supelco®
  53633-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • Description:
    Glabridin, ≥98,0% (HPLC)
  • Formula:
    C₂₀H₂₀O₄
  • Description:
    Glabridin
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.