Order Entry
France
ContactUsLinkComponent
Organic reference standard, Butyraldehyde, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabiliser, analytical standard, Supelco®
Organic reference standard, Butyraldehyde, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabiliser, analytical standard, Supelco®
Catalog # 94194-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Butyraldehyde, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabiliser, analytical standard, Supelco®
Catalog # 94194-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    77220-86-1
  • Description:
    Butyraldehyde, ≥95,0% (GC)
  • Formula:
    C₅H₁₀O₃ ·xNa⁺ ·yH₂O
  • Storage temperature:
    2...8 °C
  • Description:
    Butyraldehyde

Specifications

About this item

Butyraldehyde (BA) belongs to the class of volatile organic compounds (VOCs).
Analytical standard reference:

  • Air samples by pentafluorophenyl hydrazine derivatisation followed by analysis using gas chromatography (GC) equipped with flame ionisation detector (FID)
  • Ambient air by GC coupled to mass spectrometry (MS)
  • Saliva samples by magnetic-stirring-assisted micro-solid-phase extraction (μ-SPE) and headspace GC combined with ion-mobility spectrometry (IMS)

Also used as analytical Environmental samples by 2,4-dinitrophenylhydrazine derivatisation followed by analysis using rapid resolution liquid chromatography-ultraviolet diode array detector (RRLC-UV-DAD) and RRLC coupled to ion-trap mass spectrometer (IT-MS) operating under the atmospheric pressure chemical ionisation (APCI) mode.