Supelco® Organic Reference Standard, Betulinic acid
Catalog # 91466-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:472-15-1
- Description:Betulinic acid
- Storage conditions:2…8 °C
- Shipping conditions:WICE
- MDL No.:MFCD00009619
- M-Clarity quality level:MQ100
- Synonym(s):3β-Hydroxy-20(29)-lupaene-28-oic acid; Lupatic acid; Mairin
- Melting point:295 - 298 °C (dec.) (lit.)
- Molecular weight:456,70