Order Entry
France
ContactUsLinkComponent
Phytochemical standard, 6',7'-Epoxybergamottin, analytical standard, Supelco®
Phytochemical standard, 6',7'-Epoxybergamottin, analytical standard, Supelco®
Catalog # 69779-10MG
Supplier:  Merck
CAS Number:  
undefined
Phytochemical standard, 6',7'-Epoxybergamottin, analytical standard, Supelco®
Catalog # 69779-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
     206978-14-5
  • Description:
    6',7'-Epoxybergamottin, ≥95,0% (HPLC)
  • Formula:
    C₂₁H₂₂O₅
  • Storage temperature:
    −20 °C
  • Description:
    6',7'-Epoxybergamottin
  • Form:
    Neat

Specifications

About this item