Cetyl trimethylammonium hydrogen sulphate ≥99.0% (by titrimetric analysis) for ion pair chromatography, Supelco®
Catalog # 52371-5G-F
Supplier: Merck
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: Hexadecyltrimethylammonium Hydrogen sulphate, Cetyltrimethylammonium Hydrogen sulphate
:
:381.616 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:68214-07-3
Formula:C₁₉H₄₃NO₄S
MDL Number:MFCD00134393