Order Entry
France
ContactUsLinkComponent
Pesticide standard, Fenthion-sulfone, reference material, Honeywell Fluka™
Pesticide standard, Fenthion-sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1698-10MG
CAS Number:  
Pesticide standard, Fenthion-sulfone, reference material, Honeywell Fluka™
Catalog # HONCR1698-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    3761-42-0
  • Description:
    Fenthion-sulfone
  • Formula:
    C₁₀H₁₅O₅PS₂
  • Storage temperature:
    2-8°C
  • Description:
    Fenthion-sulfone
  • Form:
    Solid

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.