Order Entry
France
ContactUsLinkComponent
Pesticide standard, L(-)-Nicotine, reference material, Honeywell Fluka™
Pesticide standard, L(-)-Nicotine, reference material, Honeywell Fluka™
Certificates
Pesticide standard, L(-)-Nicotine, reference material, Honeywell Fluka™

Specifications

  • CAS No.:
    54-11-5
  • Formula:
    C₁₀H₁₄N₂
  • Storage temperature:
    2-8°C
  • Description:
    L(-)-Nicotine

Specifications

Frequently Bought Together