Order Entry
France
ContactUsLinkComponent
 
Pesticide standard, Diazinon, reference material, Honeywell Fluka™
Catalog # HONCR1632-100MG
CAS Number:  
Pesticide standard, Diazinon, reference material, Honeywell Fluka™
Catalog # HONCR1632-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    333-41-5
  • Description:
    Diazinon
  • Formula:
    C₁₂H₂₁N₂O₃PS
  • Storage temperature:
    2-8°C
  • Description:
    Diazinon
  • Form:
    Liquid

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.