Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, TBTC
Supelco® PESTANAL® Organic Reference Standard, TBTC
Catalog # 45713-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, TBTC
Catalog # 45713-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    1461-22-9
  • Description:
    TBTC
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H301 - H312 - H315 - H317 - H319 - H360FD - H372 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00000521
  • M-Clarity quality level:
    MQ200
  • Synonym(s):
    Tributyltin chloride; TBTC; Tributylchlorotin
  • Formula:
    [CH₃(CH₂)₃]₃SnCl
  • Molecular weight:
    325,51
  • Density:
    1,2 g/ml at 25 °C (lit.)
  • Flash point:
    108 °C
  • Boiling point:
    171 - 173 °C/25 mmHg (lit.)

Specifications

About this item

  • GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P305 + P351 + P338