Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Sulfosulfuron
Supelco® PESTANAL® Organic Reference Standard, Sulfosulfuron
  33307-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Sulfosulfuron
  33307-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    141776-32-1
  • Description:
    Sulfosulfuron
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD04034670
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-(4,6-Dimethoxypyrimidin-2-yl)-3-(2-ethylsulfonylimidazol[1,2-a]pyridin-3-ylsulfonyl)urea
  • Formula:
    C₁₆H₁₈N₆O₇S₂
  • Molecular weight:
    470,48
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS09
 : P273 - P391 - P501