Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Spinosad
Supelco® PESTANAL® Organic Reference Standard, Spinosad
Catalog # 33706-50MG
Supplier:  Merck
CAS Number:  
Supelco® PESTANAL® Organic Reference Standard, Spinosad
Catalog # 33706-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    168316-95-8
  • Description:
    Spinosad
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Mixture of Spinosyn A and Spinosyn D
  • Formula:
    C₈₃H₁₃₂N₂O₂₀

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P391 - P501