Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Quinmerac
Supelco® PESTANAL® Organic Reference Standard, Quinmerac
Catalog # 36522-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Quinmerac
Catalog # 36522-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    90717-03-6
  • Description:
    Quinmerac
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H413
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00145199
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    7-Chloro-3-methyl-8-quinolinecarboxylic acid
  • Formula:
    C₁₁H₈ClNO₂
  • Molecular weight:
    221,64
  • Flash point:
    Not applicable

Specifications

About this item

Caution: P273 - P501