Supelco® PESTANAL® Organic Reference Standard, Myclobutanil
Catalog # 34360-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:88671-89-0
- Description:Myclobutanil
- Storage conditions:2...30 °C
- Hazard class description:H302 - H319 - H361d - H411
- Shipping conditions:STD
- MDL No.:MFCD00144818
- M-Clarity quality level:MQ100
- Formula:C₁₅H₁₇ClN₄
- Molecular weight:288,78
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P202 - P264 - P273 - P301 + P312 - P305 + P351 + P338 - P308 + P313