Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Iprobenfos
Supelco® PESTANAL® Organic Reference Standard, Iprobenfos
  45814-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Iprobenfos
  45814-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    26087-47-8
  • Description:
    Iprobenfos
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00191226
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    288,34

 

 

  • GHS signal: GHS07, GHS09
 : P264 - P270 - P273 - P301 + P312 - P391 - P501