Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Fenobucarb
Supelco® PESTANAL® Organic Reference Standard, Fenobucarb
Catalog # 45488-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fenobucarb
Catalog # 45488-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    3766-81-2
  • Description:
    Fenobucarb
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00053081
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    BPMC
  • Formula:
    C₁₂H₁₇NO₂
  • Molecular weight:
    207,27
  • Flash point:
    >100 °C

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501