Supelco® PESTANAL® Organic Reference Standard, Fenarimol
45484-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:60168-88-9
- Description:Fenarimol
- Storage conditions:2...30 °C
- Hazard class description:H361fd - H362 - H411
- Shipping conditions:STD
- MDL No.:MFCD00055325
- M-Clarity quality level:MQ100
- Formula:C₁₇H₁₂Cl₂N₂O
- Molecular weight:331,20
- Flash point:100 °C - closed cup
- GHS signal: GHS08, GHS09
: P202 - P260 - P263 - P264 - P273 - P308 + P313