Order Entry
France
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Etoxazole
  32506-50MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Etoxazole
  32506-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    153233-91-1
  • Description:
    Etoxazole
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD07363987
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (RS)-5-tert-Butyl-2-[2-(2,6-difluorophenyl)-4,5-dihydro-1,3-oxazol-4-yl]phenetole; 4-(4-tert-Butyl-2-ethoxyphenyl)-2-(2,6-difluorophenyl)-4,5-dihydrooxazole
  • Formula:
    C₂₁H₂₃F₂NO₂
  • Molecular weight:
    359,41
  • Flash point:
    457 °C

 

 

  • GHS signal: GHS07, GHS09
 : P273