Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Dimoxystrobin
Supelco® PESTANAL® Organic Reference Standard, Dimoxystrobin
Catalog # 33499-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dimoxystrobin
Catalog # 33499-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    149961-52-4
  • Description:
    Dimoxystrobin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H332 - H351 - H361d - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD05664707
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (E)-2-(2,5-Dimethylphenoxymethyl)-α-methoxyimino-N-methylphenylacetamide
  • Formula:
    C₁₉H₂₂N₂O₃
  • Molecular weight:
    326,39
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P201 - P202 - P261 - P273 - P304 + P340 + P312 - P308 + P313