Supelco® PESTANAL® Organic Reference Standard, Coumaphos
Catalog # 45403-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:56-72-4
- Description:Coumaphos
- Storage conditions:2...8 °C
- Hazard class description:H300 - H311 - H410
- Shipping conditions:STD
- MDL No.:MFCD00041820
- M-Clarity quality level:MQ100
- Formula:C₁₄H₁₆ClO₅PS
- Molecular weight:362,77
- Storage temperature:2...8 °C
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P264 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P361 + P364