Order Entry
France
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Closantel
Catalog # 34093-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Closantel
Catalog # 34093-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    57808-65-8
  • Description:
    Closantel
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H301 - H413
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00661151
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5′-Chloro-4′-(4-chloro-α-cyanobenzyl)-3,5-diiodo-2′-methylsalicylanilide; N-[5-Chloro-4-(4-chloro-α-cyanobenzyl)-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide
  • Formula:
    C₂₂H₁₄Cl₂I₂N₂O₂
  • Molecular weight:
    663,07
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06
Caution: P264 - P270 - P273 - P301 + P310 - P405 - P501