Supelco® Organic Reference Standard, Cis-11-Eicosenoic Acid
44878-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:5561-99-9
- Description:cis-11-Eicosenoic acid
- Environmentally Preferable:
- Storage conditions:–20 °C
- Shipping conditions:STD
- MDL No.:MFCD00010506
- M-Clarity quality level:MQ100
- Synonym(s):Gondoic acid
- Melting point:23...24 °C (lit.)
- Formula:CH₃(CH₂)₇CH=CH(CH₂)₉CO₂H
- Molecular weight:310,51
- Storage temperature:−20 °C
- Density:0,883 g/ml at 25 °C (lit.)
- Flash point:Not applicable
: Bottomless glass bottle. Contents are inside inserted fused cone.