Supelco® PESTANAL® Organic Reference Standard, Cinmethylin
Catalog # 34237-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:87818-31-3
- Description:Cinmethylin
- Storage conditions:2...8 °C
- Hazard class description:H332 - H411
- Shipping conditions:STD
- MDL No.:MFCD01632785
- M-Clarity quality level:MQ100
- Synonym(s):exo-1-Methyl-4-(1-methylethyl)-2-[(2- methylphenyl)methoxy]-7-oxabicyclo[2.2.1]heptane
- Formula:C₁₈H₂₆O₂
- Molecular weight:274,40
- Storage temperature:2...8 °C
- Flash point:147 °C
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P271 - P273 - P304 + P340 + P312 - P391 - P501