Order Entry
France
ContactUsLinkComponent
Pesticide reference standard, Chlorpyrifos solution, PESTANAL®, analytical standard, Supelco®
Pesticide reference standard, Chlorpyrifos solution, PESTANAL®, analytical standard, Supelco®
Catalog # 31553-10ML
Supplier:  Merck
CAS Number:  
Pesticide reference standard, Chlorpyrifos solution, PESTANAL®, analytical standard, Supelco®
Catalog # 31553-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    10 mL
  • Standards:
    Analytical standard
  • CAS No.:
    2921-88-2
  • Description:
    Chlorpyrifos solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₉H₁₁Cl₃NO₃PS
  • Storage temperature:
    2...8 °C
  • Description:
    Chlorpyrifos solution

Specifications

About this item