Order Entry
France
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Carboxine
Supelco® PESTANAL® Organic Reference Standard, Carboxine
Catalog # 45371-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Carboxine
Catalog # 45371-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    5234-68-4
  • Description:
    Carboxine
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H317 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055403
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5,6-Dihydro-2-methyl-N-phenyl-1,4-oxathiin-3-carboxamide; Carbathiine
  • Formula:
    C₁₂H₁₃NO₂S
  • Molecular weight:
    235,30
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P260 - P273 - P280 - P301 + P312 - P302 + P352 - P314