Supelco® PESTANAL® Organic Reference Standard, Carbofuran-D₃
34019-10MG-R
: Merck
:
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1007459-98-4
- Description:Carbofuran-d₃
- Storage conditions:2...30 °C
- Hazard class description:H300 + H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD07366666
- M-Clarity quality level:MQ100
- Synonym(s):2,3-Dihydro-2,2-dimethyl-7-benzofuranol N-methylcarbamate-d₃
- Formula:C₁₂H₁₂D₃NO₃
- Molecular weight:224,27
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P260 - P264 - P270 - P271 - P273 - P304 + P340 + P310